Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2R)-(-)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct ≥97%

Supplier: STREM CHEMICALS INC MS

44-0210-50MG 44-0210-10MG 44-0210-250MG
100199-886EA 400.55 USD
100199-886 100206-634 100199-888
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2R)-(-)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct ≥97%
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(2R)-(-)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) dichloromethane adduct

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD02684563
CAS Number: 329735-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR