Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tetrachlorobis(2,7-dimethyl-2,6-octadienylene)diruthenium, Dichlorobis(μ-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV), Dichlorodi-μ-chlorobis[(1,2,3,6,7,8-η-2,7-dimethyl-2,6-octadiene-1,8-diyl]diruthenium(IV)

44-0203-1G 44-0203-250MG
101418-926EA 369.42 USD
101418-926 101418-924
Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV) ≥99%
Dichlorobis(mu-chloro)bis[(1,2,3,6,7,8-η)-2,7-dimethyl-2,6-octadien-1,8-diyl]diruthenium(IV)

Color and Form: purple pwdr.
Stability: moisture sensitive

Formula: C₂₀H₃₂Cl₄Ru₂
Melting Pt: 196 °C (dec.)
MDL Number: MFCD08459355
CAS Number: 34801-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR