Dichloro(p-cymene)ruthenium(II) dimer ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

44-0190-5G 44-0190-1G
100201-640EA 291.57 USD
100201-640 100207-682
Dichloro(p-cymene)ruthenium(II) dimer ≥98%
Dichloro(p-cymene)ruthenium(II) dimer
Formula: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
MDL Number: MFCD00064793
CAS Number: 52462-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR