Dicarbonylcyclopentadienylruthenium dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

44-0175-2G 44-0175-500MG
100195-932EA 1126.65 USD
100195-932 100208-434
Dicarbonylcyclopentadienylruthenium dimer ≥99%
Dicarbonylcyclopentadienylruthenium dimer

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₄H₁₀O₄Ru₂
MDL Number: MFCD00070447
CAS Number: 12132-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR