Benzeneruthenium(II) chloride dimer ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dichloro(benzene)ruthenium(II) dimer

44-0170-250MG 44-0170-1G
100208-896EA 103.32 USD
100208-896 100208-898
Benzeneruthenium(II) chloride dimer ≥98%
Benzeneruthenium(II) chloride dimer
Formula: C₁₂H₁₂Cl₄Ru₂
MDL Number: MFCD00064686
CAS Number: 37366-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR