Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ru(OAc)2[(R)-H8-binap]

44-0166-50MG 44-0166-250MG
101418-896EA 117.48 USD
101418-896 101418-898
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl]ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753023
CAS Number: 374067-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR