Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ru(OAc)2[(S)-tolbinap]

44-0163-250MG 44-0163-1G
101418-884EA 75.02 USD
101418-884 101418-886
Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753021
CAS Number: 106681-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR