Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ru(OAc)2[(R)-tolbinap]

44-0162-250MG 44-0162-1G
101418-880EA 75.02 USD
101418-880 101418-882
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753021
CAS Number: 116128-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR