Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cyclopentadienyl(p-cymene) ruthenium(II)hexafluorophosphate

44-0160-250MG 44-0160-1G
100203-152EA 206.65 USD
100203-152 100203-154
Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate ≥98%
Cyclopentadienyl(p-cymene)ruthenium(II) hexafluorophosphate

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₅H₁₉F₆PRu
MW: 445.3500 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD06798307
CAS Number: 147831-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR