Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-Ru(OAc)2(BINAP), Ru(OAc)2[(S)-binap]
44-0153-1G
44-0153-250MG
101418-878EA
223.63
USD
101418-878
101418-876
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
BINAP
Formula:
C₄₈H₃₈O₄P₂Ru MW: 841.83 g/mol Melting Pt: >100 °C (dec.) |
MDL Number:
MFCD09753020 CAS Number: 261948-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...