RuCl[(R,R)-Tsdpen](p-cymene) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), RuCl(p-cymene)[(R,R)-Ts-DPEN], Chloro(p-cymene)[(R,R)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), Chloro{[(1R,2R)-(-)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II)

44-0148-1G 44-0148-250MG
101442-328EA 223.63 USD
101442-328 101442-326
RuCl[(R,R)-Tsdpen](p-cymene) ≥95%
RuCl[(R,R)-Tsdpen](p-cymene)
Formula: C₃₁H₃₅ClN₂O₂RuS
MW: 636.21 g/mol
MDL Number: MFCD10567385
CAS Number: 192139-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR