Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0145-2G 44-0145-500MG
100196-948EA 1295.08 USD
100196-948 100200-978
Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer ≥95%
Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer

Metallocenes, Derivatives & Cp Precursors

Formula: C₄₀H₆₀Ru₄Cl₄
MW: 1087.0000 g/mol
MDL Number: MFCD03792150
CAS Number: 113860-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR