Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro(indenyl)bis(triphenylphosphine)ruthenium(II)

44-0125-5G 44-0125-1G
100201-638EA 502.46 USD
100201-638 100201-636
Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct ≥98%
Chloro(indenyl)bis(triphenylphosphine)ruthenium(II),dichloromethane adduct

Metallocenes, Derivatives & Cp Precursors

Formula: C₄₅H₃₇ClP₂Ru
MW: 776.2500 g/mol
MDL Number: MFCD04113601
CAS Number: 99897-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR