Chlorodicarbonyl[1-(i-propylamino)-2,3,4,5-tetraphenylcyclopentadienyl]ruthenium(II) ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

44-0123-2G 44-0123-500MG
100205-946EA 1259.69 USD
100205-946 100200-272
Chlorodicarbonyl[1-(i-propylamino)-2,3,4,5-tetraphenylcyclopentadienyl]ruthenium(II) ≥95%
Chlorodicarbonyl[1-(i-propylamino)-2,3,4,5-tetraphenylcyclopentadienyl]ruthenium(II)

Metallocenes, Derivatives & Cp Precursors

Melting Pt: 197 °C (dec.)
MDL Number: MFCD04038739
CAS Number: 470688-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR