Chloro(cyclopentadienyl)[bis(diphenylphosphino)methane]ruthenium(II) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

44-0116-250MG 44-0116-1G
101324-716EA 382.15 USD
101324-716 101324-718
Chloro(cyclopentadienyl)[bis(diphenylphosphino)methane]ruthenium(II) ≥97%
Chloro(cyclopentadienyl)[bis(diphenylphosphino)methane]ruthenium(II)

Metallocenes, Derivatives & Cp Precursors

Formula: C₃₀H₂₄ClP₂Ru
MDL Number: MFCD07782002
CAS Number: 71397-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR