Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-0100-1G
44-0100-250MG
100198-764EA
253.35
USD
100198-764
100197-514
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)
Formula:
C₅₅H₄₇OP₃Ru MW: 917.97 g/mol |
MDL Number:
MFCD00015870 CAS Number: 25360-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...