Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

Supplier: STREM CHEMICALS INC MS
Warning

44-0094-50MG 44-0094-250MG
101418-840EA 97.66 USD
101418-840 101418-842
Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride
Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

BINAP

Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753016
CAS Number: 944451-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR