Carbonylbis(trifluoroacetato)bis(triphenylphosphine)ruthenium(II) methanol adduct ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

44-0090-1G 44-0090-250MG
100198-762EA 672.31 USD
100198-762 100209-940
Carbonylbis(trifluoroacetato)bis(triphenylphosphine)ruthenium(II) methanol adduct ≥98%
Carbonylbis(trifluoroacetato)bis(triphenylphosphine)ruthenium(II) methanol adduct
Melting Pt: 230 °C (dec.)
MDL Number: MFCD06658132
CAS Number: 38596-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR