[RuCl(p-cymene)((S)-tolbinap)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, Chloro[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-tolbinap)]Cl

44-0089-250MG 44-0089-1G
101418-828EA 75.02 USD
101418-828 101418-830
[RuCl(p-cymene)((S)-tolbinap)]Cl
[RuCl(p-cymene)((S)-tolbinap)]Cl

BINAP

Formula: C₅₈H₅₄Cl₂P₂Ru
MW: 984.99 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753014
CAS Number: 228120-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR