Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

Supplier: STREM CHEMICALS INC MS
Warning

44-0088-1G 44-0088-250MG
101418-826EA 223.63 USD
101418-826 101418-824
Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride
Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

BINAP

Formula: C₅₈H₅₄ClP₂RuCl
MW: 984.9900 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD09753014
CAS Number: 131614-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR