[RuCl(p-cymene)((S)-binap)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

44-0086-250MG 44-0086-1G
101418-820EA 75.02 USD
101418-820 101418-822
[RuCl(p-cymene)((S)-binap)]Cl
[RuCl(p-cymene)((S)-binap)]Cl

BINAP

Formula: C₅₄H₄₆Cl₄P₂Ru
Melting Pt: >100 °C (dec.)
MDL Number: MFCD00134456
CAS Number: 130004-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR