Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

44-0085-1G 44-0085-5G
100200-466EA 118.89 USD
100200-466 100200-468
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) ≥99%
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Formula: C₅₅H₄₅ClOP₃Ru
MW: 951.405 g/mol
Melting Pt: 209…210 °C
MDL Number: MFCD00049804
CAS Number: 16971-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR