[RuCl(p-cymene)((R)-binap)]Cl

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-binap)]Cl, (R)-RuCl[(p-cymene)(BINAP)]Cl, Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(R)-Binap RuCl p-cymene]Cl

44-0084-250MG 44-0084-1G
101418-816EA 75.02 USD
101418-816 101418-818
[RuCl(p-cymene)((R)-binap)]Cl
[RuCl(p-cymene)((R)-binap)]Cl

BINAP

Formula: C₅₄H₄₆Cl₂P₂Ru
MW: 928.88 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD00134456
CAS Number: 145926-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR