Bis(tricyclohexylphosphine)[(phenylthio)methylene]ruthenium(II) dichloride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

44-0073-1G 44-0073-5G
100200-462EA 155.69 USD
100200-462 100200-464
Bis(tricyclohexylphosphine)[(phenylthio)methylene]ruthenium(II) dichloride ≥97%
Bis(tricyclohexylphosphine)[(phenylthio)methylene]ruthenium(II) dichloride

Metathesis Catalyst

MDL Number: MFCD06658131
CAS Number: 219770-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR