(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(tricyclohexylphosphine)-3-phenyl-1H-inden-1-ylideneruthenium(II) dichloride

44-0063-5G 44-0063-1G
101324-702EA 585.97 USD
101324-702 101324-700
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran

Metathesis Catalyst

Formula: C₅₁H₇₆Cl₂P₂Ru
MW: 923.09 g/mol
MDL Number: MFCD07369034
CAS Number: 250220-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR