Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II) ≥99% (99.9%-Ru)

Supplier: STREM CHEMICALS INC MS
Warning

44-0060-5G 44-0060-1G
100204-056EA 914.34 USD
100204-056 100204-054
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II) ≥99% (99.9%-Ru)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)(1,5-cyclooctadiene)ruthenium(II)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₀H₅₀O₄Ru
Boiling Pt: 220 °C/ 100 °C (0,05 mmHg) (dec./ subl.)
Melting Pt: 187…190 °C
MDL Number: MFCD01073791
CAS Number: 329735-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR