Bis(pentamethylcyclopentadienyl)ruthenium(II) ≥99% (99.9%-Ru)

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(pentamethylcyclopentadienyl)ruthenium, Bis(pentamethylcyclopentadienyl)ruthenium(II), Decamethylruthenocene

44-0050-500MG 44-0050-2G
100208-894EA 151.45 USD
100208-894 100205-944
Bis(pentamethylcyclopentadienyl)ruthenium(II) ≥99% (99.9%-Ru)
Bis(pentamethylcyclopentadienyl)ruthenium (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₀H₃₀Ru
MW: 371.522 g/mol
MDL Number: MFCD00058889
CAS Number: 84821-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR