Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium (II) complex, Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), Bis(2-methylallyl)-1,5-cyclooctadieneruthenium (II)
44-0046-250MG
44-0046-1G
100201-634EA
77.85
USD
100201-634
100206-624
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) ≥97%
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)
Volatile Precursors for CVD
Formula:
C₁₆H₂₆Ru Melting Pt: 80…85 °C |
MDL Number:
MFCD00216965 CAS Number: 12289-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...