Bis(ethylcyclopentadienyl)ruthenium(II) ≥98% (99.9%-Ru)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
44-0040-2G
44-0040-500MG
100198-760EA
1027.57
USD
100198-760
100209-938
Bis(ethylcyclopentadienyl)ruthenium(II) ≥98% (99.9%-Ru)
Bis(ethylcyclopentadienyl)ruthenium(II)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Density:
1,3412 |
MDL Number:
MFCD03427334 CAS Number: 32992-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...