Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

44-0015-1G 44-0015-250MG
101324-698EA 835.08 USD
101324-698 101324-696
Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate ≥98%
Acetonitrilebis[2-diphenylphosphino-6-t-butylpyridine]cyclopentadienylruthenium(II) hexafluorophosphate

Metallocenes, Derivatives & Cp Precursors

Formula: C₄₉H₅₂F₆N₃P₃Ru
MDL Number: MFCD06798306
CAS Number: 776230-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR