Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

42-2200-1G 42-2200-5G
100197-512EA 131.63 USD
100197-512 100199-884
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer ≥99%
Pentamethylcyclopentadienylmolybdenum dicarbonyl dimer

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₄H₃₀Mo₂O₄
Melting Pt: 219 °C (dec.)
MDL Number: MFCD00058846
CAS Number: 12132-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR