Molybdenum(IV) 2-ethylhexanoate (15% Mo)

Supplier: STREM CHEMICALS INC MS
Warning

42-1400-500G 42-1400-100G
100208-892EA 2679.32 USD
100208-892 100207-676
Molybdenum(IV) 2-ethylhexanoate (15% Mo)
Molybdenum(IV) 2-ethylhexanoate
Density: 1,11…1.15
Flash Pt: 250 °F
MDL Number: MFCD01631280
CAS Number: 34041-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR