Molybdenum(VI) dioxide bis(2,2,6,6-tetramethyl-3,5-heptanedionate) ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: [O2Mo(TMHD)2]

42-1357-1G 42-1357-5G 42-1357-25G
100205-124EA 147.2 USD
100205-124 100209-934 100203-142
Molybdenum(VI) dioxide bis(2,2,6,6-tetramethyl-3,5-heptanedionate) ≥98%
Molybdenum(VI) dioxide bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

Metal TMHD

Melting Pt: 129…131 °C
MDL Number: MFCD00269827
CAS Number: 34872-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR