2,6-Diisopropylphenylimidoneophylidene[racemic-BIPHEN]molybdenum(VI) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

42-1212-100MG 42-1212-500MG
100203-138EA 240.62 USD
100203-138 100203-140
2,6-Diisopropylphenylimidoneophylidene[racemic-BIPHEN]molybdenum(VI) ≥97%
2,6-Diisopropylphenylimidoneophylidene[racemic-BIPHEN]molybdenum(VI)

Schrock-Hoveyda Catalyst

MDL Number: MFCD01862462
CAS Number: 300344-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR