2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide)

Supplier: STREM CHEMICALS INC MS

42-1205-2G 42-1205-500MG 42-1205-100MG
100200-968EA 2936.92 USD
100200-968 100203-136 100209-932
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide)
2,6-Diisopropylphenylimidoneophylidene molybdenum(VI) bis(hexafluoro-t-butoxide)

Alkoxide, Schrock's Catalyst

MDL Number: MFCD00269857
CAS Number: 139220-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR