Ammonium tetrathiomolybdate ≥99% (99.99%-Mo), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ammonium tetrathiomolybdate(VI)

42-1000-25G 42-1000-5G
100197-022EA 815.26 USD
100197-022 100206-612
Ammonium tetrathiomolybdate ≥99% (99.99%-Mo), PURATREM
Ammonium tetrathiomolybdate
Formula: H₁₀MoN₂S₄
MW: 262.284 g/mol
MDL Number: MFCD00136013
CAS Number: 15060-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR