Cyclopentadienylmolybdenum(V) tetrachloride ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

42-0390-5G 42-0390-1G
100210-644EA 564.74 USD
100210-644 100206-610
Cyclopentadienylmolybdenum(V) tetrachloride ≥95%
Cyclopentadienylmolybdenum(V) tetrachloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₅H₅Cl₄Mo
MW: 302.845 g/mol
MDL Number: MFCD00269826
CAS Number: 62927-99-5
UN: 1759
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR