(Cycloheptatriene)molybdenum tricarbonyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

42-0350-5G 42-0350-1G
100198-182EA 741.66 USD
100198-182 100200-966
(Cycloheptatriene)molybdenum tricarbonyl ≥99%
(Cycloheptatriene)molybdenum tricarbonyl

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₈MoO₃
Melting Pt: 100…101 °C
MDL Number: MFCD00004160
CAS Number: 12125-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR