Bis(cyclopentadienyl)molybdenum dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

42-0100-1G 42-0100-5G
100196-212EA 230.71 USD
100196-212 100196-214
Bis(cyclopentadienyl)molybdenum dichloride ≥99%
Bis(cyclopentadienyl)molybdenum dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₀Cl₂Mo
MDL Number: MFCD00009948
CAS Number: 12184-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR