Niobium(IV) 2-ethylhexanoate

Supplier: STREM CHEMICALS INC MS
Warning

41-1260-50G 41-1260-10G
100206-606EA 1222.89 USD
100206-606 100206-604
Niobium(IV) 2-ethylhexanoate
Niobium(IV) 2-ethylhexanoate
MDL Number: MFCD01631279
CAS Number: 206564-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR