Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), Zr(TMHD)4

40-5000-25G 40-5000-1G 40-5000-5G
100201-222EA 675.14 USD
100201-222 100197-506 100204-048
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV) ≥99%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₄₄H₇₆O₈Zr
MW: 824.29 g/mol
Boiling Pt: 180 °C (0.1 mmHg) (subl.)
Melting Pt: 318…320 °C
MDL Number: MFCD00145380
CAS Number: 18865-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR