Zirconium disulphate tetrahydrate ≥99.99% (Zr basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

40-4750-100G 40-4750-25G
100205-942EA 830.83 USD
100205-942 100196-208
Zirconium disulphate tetrahydrate ≥99.99% (Zr basis), PURATREM
Zirconium disulphate tetrahydrate
Formula: O₈S₂Zr·4H₂O
MW: 355.396 g/mol
Melting Pt: 100 °C (-3H₂O)
MDL Number: MFCD00149267
CAS Number: 7446-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR