Tetrakis(dimethylamino)zirconium(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
TDMAZ, Tetrakis(dimethylamino)zirconium
40-4100-1G
40-4100-5G
40-4100-25G
100208-422EA
118.89
USD
100208-422
100201-624
100203-120
Tetrakis(dimethylamino)zirconium(IV) ≥99%
Tetrakis(dimethylamino)zirconium(IV)
Metal Alkylamides, Metallocenes, Derivatives & Cp Precursors, Volatile Precursors for CVD
Formula:
C₈H₂₄N₄Zr MW: 267.527 g/mol Boiling Pt: 80 °C (0.1 mmHg) Melting Pt: 57…60 °C |
MDL Number:
MFCD00239502 CAS Number: 19756-04-8 UN: 3131 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...