Zirconium(IV) isopropoxide isopropanol complex ≥99.9% (Zr basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zirconium(IV) i-propoxide (isopropanol adduct)

40-4016-50G 40-4016-10G
100203-782EA 760.06 USD
100203-782 100209-344
Zirconium(IV) isopropoxide isopropanol complex ≥99.9% (Zr basis)
Zirconium(IV) isopropoxide isopropanol complex

Alkoxide

Formula: C₁₅H₃₆O₅Zr
MW: 387.672 g/mol
Boiling Pt: Dec.
MDL Number: MFCD00075157
CAS Number: 14717-56-7
UN: 1325
ADR: 4 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR