Zirconium(IV) oxide-yttria ≥99% stabilized

Supplier: STREM CHEMICALS INC MS

40-4002-100G 40-4002-500G
100199-876EA 249.11 USD
100199-876 100199-878
Zirconium(IV) oxide-yttria ≥99% stabilized
Zirconium(IV) oxide-yttria
MDL Number: MFCD00145546
CAS Number: 64417-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR