Zirconium(IV) oxide-yttria ≥99% stabilised

Supplier: STREM CHEMICALS INC MS

40-4000-500G 40-4000-100G
100201-220EA 1051.63 USD
100201-220 100196-204
Zirconium(IV) oxide-yttria ≥99% stabilised
Zirconium(IV) oxide-yttria
MDL Number: MFCD00145546
CAS Number: 64417-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR