Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Zirconyl 2-ethylhexanoate

40-2750-500G 40-2750-100G
100200-960EA 165.6 USD
100200-960 100209-920
Zirconium(IV) oxide-2-ethylhexanoate (∼6% Zr) in mineral spirits
Zirconium(IV) oxide-2-ethylhexanoate
Formula: (C₈H₁₅O₂)xZn
Flash Pt: 104 °F
MDL Number: MFCD00072684
CAS Number: 22464-99-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR