Tetrachlorobis(tetrahydrofuran)zirconium(IV)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetrachlorobis(tetrahydrofuran)zirconium

40-1700-10G 40-1700-50G
100201-620EA 150.03 USD
100201-620 100205-938
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
Formula: C₄H₈Cl₄OZr
MW: 377.25 g/mol
MDL Number: MFCD00145367
CAS Number: 21959-01-3
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR