(Pentamethylcyclopentadienyl)zirconium trichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

40-1500-5G 40-1500-1G
100201-618EA 549.17 USD
100201-618 100201-616
(Pentamethylcyclopentadienyl)zirconium trichloride ≥99%
(Pentamethylcyclopentadienyl)zirconium trichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₅Cl₃Zr
MW: 332.81 g/mol
MDL Number: MFCD00070444
CAS Number: 75181-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR