Dimethylbis(indenyl)zirconium ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

40-1120-5G 40-1120-1G
100201-216EA 716.18 USD
100201-216 100200-950
Dimethylbis(indenyl)zirconium ≥98%
Dimethylbis(indenyl)zirconium

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD01074449
CAS Number: 49596-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR